siRNA Details
Gene Symbol: UROD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF104421.1 | - | 1284 | |||
AF104422.1 | - | 1284 | ||||
AF104423.1 | - | 1284 | ||||
AF104424.1 | - | 1284 | ||||
AF104425.1 | - | 1284 | ||||
AF104426.1 | - | 1284 | ||||
AF104427.1 | - | 1284 | ||||
AF104428.1 | - | 1284 | ||||
AF104429.1 | - | 1284 | ||||
AF104430.1 | - | 1284 | ||||
AF104431.1 | - | 1284 | ||||
AF104432.1 | - | 1284 | ||||
AF104433.1 | - | 1284 | ||||
AF104434.1 | - | 1284 | ||||
AF104435.1 | - | 1284 | ||||
AF104436.1 | - | 1284 | ||||
AF104437.1 | - | 1284 | ||||
AF104438.1 | - | 1284 | ||||
AF104439.1 | - | 1284 | ||||
AF104440.1 | - | 1284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
