siRNA Details
Gene Symbol: MYH7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000947.1 | - | 5 | 604 | ||
BC069327.1 | - | 5 | 587 | |||
BC069406.1 | - | 5 | 587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
