siRNA Details
Gene Symbol: M6PR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207024.1 | NP_001193953.1 | 5 | 1773 | ||
NM_002355.3 | NP_002346.1 | 7 | 2031 | |||
XM_005253376.2 | XP_005253433.1 | 7 | 1893 | |||
GenBank mRNA | BC017111.1 | - | 5 | 1431 | ||
BC024206.1 | - | 7 | 1923 | 3530887 | ||
BG750588.1 | - | 1 | 405 | |||
M16985.1 | - | 7 | 1901 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
