siRNA Details
Gene Symbol: LILRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081442.2 | NP_001074911.1 | 9, 10 | 1589 | ||
NM_001081443.2 | NP_001074912.1 | 8, 9 | 1289 | |||
NM_001304457.1 | NP_001291386.1 | 9, 10 | 1562 | |||
NM_006840.4 | NP_006831.1 | 9, 10 | 1586 | |||
XM_011526359.2 | XP_011524661.1 | 9, 10 | 1573 | |||
XM_011526360.2 | XP_011524662.1 | 9, 10 | 1570 | |||
XM_011526361.2 | XP_011524663.1 | 8, 9 | 1522 | |||
XM_011526362.2 | XP_011524664.1 | 8, 9 | 1254 | |||
GenBank mRNA | AF025534.1 | - | 9, 10 | 1537 | ||
AK057072.1 | - | 8, 9 | 1243 | |||
AK092867.1 | - | 8, 9 | 2487 | |||
AK096122.1 | - | 5, 6 | 1962 | |||
AK223296.1 | - | 9, 10 | 1545 | |||
AK290845.1 | - | 9, 10 | 1570 | |||
AK301112.1 | - | 9, 10 | 1525 | |||
BC025704.1 | - | 9, 10 | 1540 | 5223581 |
