siRNA Details
Gene Symbol: DDX19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014449.2 | NP_001014449.1 | 3 | 407 | ||
NM_001257173.1 | NP_001244102.1 | 4 | 499 | |||
NM_001257174.1 | NP_001244103.1 | 3 | 453 | |||
NM_007242.5 | NP_009173.1 | 5 | 510 | |||
XM_006721127.2 | XP_006721190.1 | 4 | 432 | |||
XM_011522832.2 | XP_011521134.1 | 6 | 566 | |||
XM_011522834.2 | XP_011521136.1 | 5 | 455 | |||
XM_017022890.1 | XP_016878379.1 | 5 | 548 | |||
GenBank mRNA | AF353720.1 | - | 3 | 453 | ||
AJ237946.1 | - | 5 | 371 | |||
AK027378.1 | - | 5 | 475 | |||
AK223115.1 | - | 5 | 481 | |||
AK301938.1 | - | 4 | 499 | |||
AK316346.1 | - | 3 | 388 | |||
BC003626.2 | - | 5 | 474 | 3030038 | ||
BC010008.2 | - | 3 | 359 | 4298615 | ||
BC110805.1 | - | 4 | 281 | |||
CR457215.1 | - | 5 | 371 | |||
DC385044.1 | - | 5 | 513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
