siRNA Details
Gene Symbol: HMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015980.4 | NP_057064.1 | 5 | 1009 | ||
GenBank mRNA | AF113537.1 | - | 5 | 877 | ||
AF133422.1 | - | 5 | 861 | |||
AK098398.1 | - | 1417 | ||||
AK294383.1 | - | 4 | 792 | |||
AK295206.1 | - | 4 | 692 | |||
AK297196.1 | - | 5 | 815 | |||
AK315949.1 | - | 3 | 624 | |||
AK315952.1 | - | 4 | 706 | |||
BC002619.2 | - | 5 | 892 | 3160345 | ||
BC018873.2 | - | 5 | 885 | |||
CD627354.1 | - | 1 | 62 |
