siRNA Details
Gene Symbol: ORM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000607.2 | NP_000598.2 | 5, 6 | 646 | ||
GenBank mRNA | AK312035.1 | - | 5, 6 | 571 | ||
BC026238.1 | - | 5, 6 | 571 | 4767956 | ||
BC104818.1 | - | 5, 6 | 560 | 8143821 | ||
BC104820.1 | - | 5, 6 | 560 | 8143823 | ||
BC143313.1 | - | 5, 6 | 560 | 9051818 | ||
BT019790.1 | - | 5, 6 | 535 | |||
EU794584.1 | - | 5, 6 | 535 | |||
M13692.1 | - | 5, 6 | 550 | |||
X02544.1 | - | 5, 6 | 613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
