siRNA Details
Gene Symbol: SCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007098.2 | NP_001007099.1 | 9 | 977 | ||
NM_001193599.1 | NP_001180528.1 | 9 | 1037 | |||
NM_001193600.1 | NP_001180529.1 | 9 | 977 | |||
NM_001193617.1 | NP_001180546.1 | 9 | 1051 | |||
NM_002979.4 | NP_002970.2 | 10 | 1109 | |||
XM_005271103.4 | XP_005271160.1 | 10 | 1052 | |||
XM_011541935.2 | XP_011540237.1 | 10 | 1052 | |||
XM_017002045.1 | XP_016857534.1 | 10 | 1052 | |||
GenBank mRNA | AB208789.1 | - | 10 | 1061 | ||
AK294631.1 | - | 9 | 980 | |||
AK295214.1 | - | 9 | 994 | |||
AK308105.1 | - | 9 | 919 | |||
AK312856.1 | - | 10 | 1060 | |||
BC067108.1 | - | 9 | 888 | 5176900 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
