siRNA Details
Gene Symbol: OBP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014582.2 | NP_055397.1 | 4, 5 | 431 | ||
GenBank mRNA | AJ251021.1 | - | 4, 5 | 431 | ||
AM087898.1 | - | 4, 5 | 347 | |||
BC069563.1 | - | 4, 5 | 416 | 7262146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
