siRNA Details
Gene Symbol: Selenbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009150.3 | NP_033176.2 | 6 | 663 | ||
GenBank mRNA | AK002447.1 | - | 6 | 681 | ||
AK218745.1 | - | 2 | 191 | |||
BC011202.1 | - | 6 | 663 | 4209393 | ||
M32032.1 | - | 646 |
Gene Symbol: Selenbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019414.2 | NP_062287.1 | 6 | 673 | ||
GenBank mRNA | BC024106.1 | - | 6 | 671 | 5096831 | |
S56599.1 | - | 6 | 673 |

More Information
Other Species Targeted

Gene Ontology Categories:
