siRNA Details
Gene Symbol: Selenbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019414.2 | NP_062287.1 | 10 | 1192 | ||
GenBank mRNA | BC024106.1 | - | 10 | 1190 | 5096831 | |
S56599.1 | - | 10 | 1192 |
Gene Symbol: Selenbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009150.3 | NP_033176.2 | 10 | 1182 | ||
GenBank mRNA | AK002447.1 | - | 10 | 1198 | ||
BC011202.1 | - | 10 | 1182 | 4209393 | ||
M32032.1 | - | 1165 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
