siRNA Details
Gene Symbol: SMG1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135305.1 | 14 | 2095 | |||
NR_135317.1 | 12 | 2290 | ||||
GenBank mRNA | AB055226.1 | - | 14 | 2506 | ||
BX640895.1 | - | 2 | 2844 | |||
DA936903.1 | - | 3 | 487 |
Gene Symbol: BOLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK311534.1 | - | 3 | 487 |
Gene Symbol: BOLA2-SMG1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320622.1 | NP_001307551.1 | 5 | 1465 | ||
NM_001320623.1 | NP_001307552.1 | 5 | 1399 | |||
NM_001320624.1 | NP_001307553.1 | 4 | 1405 | |||
NM_001320625.1 | NP_001307554.1 | 4 | 1301 | |||
NM_001320627.1 | NP_001307556.1 | 5 | 1503 | |||
NM_001320628.1 | NP_001307557.1 | 4 | 821 | |||
NR_135316.1 | 4 | 1278 | ||||
GenBank mRNA | AF271775.1 | - | 6 | 1124 | ||
AK027255.1 | - | 6 | 2967 | |||
AK295153.1 | - | 5 | 807 | |||
BC062756.1 | - | 5 | 884 | |||
BG282633.1 | - | 3 | 548 | |||
BI517472.1 | - | 4 | 628 |
Gene Symbol: SMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023067.1 | XP_016878556.1 | 1 | 20 | ||
GenBank mRNA | BE079340.1 | - | 2 | 508 |
Gene Symbol: SMG1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002453.5 | 13 | 2231 | |||
GenBank mRNA | BC063703.1 | - | 13 | 2231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
