siRNA Details
Gene Symbol: UBE2S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014501.2 | NP_055316.2 | 2 | 472 | ||
XM_011526752.1 | XP_011525054.1 | 2 | 661 | |||
GenBank mRNA | AB062397.1 | - | 1 | 282 | ||
BC004236.2 | - | 1 | 282 | 2988449 | ||
BC007554.1 | - | 1 | 282 | 2967547 | ||
BC065364.1 | - | 1 | 292 | 5434942 | ||
BC066948.1 | - | 1 | 128 | 4836851 | ||
M91670.1 | - | 2 | 163 |
