siRNA Details
Gene Symbol: Trib3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175093.2 | NP_780302.2 | 4 | 1970 | ||
GenBank mRNA | AK089931.1 | - | 4 | 1848 | ||
AK132257.1 | - | 4 | 1909 | |||
AK144752.1 | - | 4 | 1889 | |||
AK146340.1 | - | 4 | 1899 | |||
AK146421.1 | - | 4 | 1827 | |||
AK159760.1 | - | 4 | 1910 | |||
AK160099.1 | - | 4 | 1960 | |||
AK195825.1 | - | 1 | 107 | |||
AK197033.1 | - | 1 | 107 | |||
AK200016.1 | - | 1 | 107 | |||
BC012955.1 | - | 5 | 1864 | 3980838 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
