siRNA Details
Gene Symbol: Trib3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175093.2 | NP_780302.2 | 4 | 2040 | ||
GenBank mRNA | AK132257.1 | - | 4 | 1979 | ||
AK144752.1 | - | 4 | 1959 | |||
AK146340.1 | - | 4 | 1969 | |||
AK200016.1 | - | 1 | 177 | |||
BC012955.1 | - | 5 | 1934 | 3980838 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
