siRNA Details
Gene Symbol: DLEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002605.2 | 2 | 690 | |||
NR_109973.1 | 3 | 743 | ||||
GenBank mRNA | AJ412023.1 | - | LIT1929 BCMS |
2 | 688 | |
AJ412024.1 | - | LIT1928 BCMS |
3 | 741 | ||
AK095311.1 | - | 3 | 663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
