siRNA Details
Gene Symbol: POM121L9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003714.1 | 7 | 3314 | |||
GenBank mRNA | AL117401.1 | - | 7 | 3108 | ||
AL117484.1 | - | 4 | 1510 | |||
BC063879.1 | - | 2 | 811 | |||
BC086868.1 | - | 3 | 1636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
