siRNA Details
Gene Symbol: FBLIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024216.1 | NP_001019387.1 | 5 | 1151 | ||
NM_017556.2 | NP_060026.2 | 9 | 1721 | |||
XM_005245909.3 | XP_005245966.1 | 10 | 2033 | |||
XM_011541617.2 | XP_011539919.1 | 10 | 1932 | |||
XM_011541618.2 | XP_011539920.1 | 9 | 1814 | |||
XM_017001525.1 | XP_016857014.1 | 10 | 1955 | |||
GenBank mRNA | AK027444.1 | - | 5 | 1169 | ||
AK055259.1 | - | 9 | 2062 | |||
AL133035.1 | - | 5 | 904 | |||
BC019895.1 | - | 10 | 1865 | 5139869 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
