siRNA Details
Gene Symbol: KRT8P41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027713.1 | 1 | 1025 | |||
GenBank mRNA | AK127993.1 | - | 1 | 1025 | ||
BC093663.1 | - | 1 | 326 | |||
BC112006.1 | - | 1 | 326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
