siRNA Details
Gene Symbol: OR6N1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005185.1 | NP_001005185.1 | 1 | 533 | ||
XM_017000325.1 | XP_016855814.1 | 3 | 1391 | |||
XM_017000326.1 | XP_016855815.1 | 4 | 1972 | |||
XM_017000327.1 | XP_016855816.1 | 3 | 765 | |||
GenBank mRNA | BC146847.1 | - | 1 | 553 | 9056775 |
