siRNA Details
Gene Symbol: VWA5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130142.1 | NP_001123614.1 | 11, 12 | 1461 | ||
NM_014622.4 | NP_055437.2 | 10, 11 | 1346 | |||
XM_011542828.2 | XP_011541130.1 | 11, 12 | 1572 | |||
GenBank mRNA | AF002672.1 | - | 9, 10 | 1258 | ||
AK291698.1 | - | 10, 11 | 1300 | |||
AK314934.1 | - | 3, 4 | 311 | |||
AY366505.1 | - | 10, 11 | 1346 | |||
AY366508.1 | - | 11, 12 | 1461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
