siRNA Details
Gene Symbol: KRTAP22-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181620.1 | NP_853651.1 | 1 | 20 | ||
GenBank mRNA | AB096953.1 | - | 1 | 54 | ||
BC101682.1 | - | 1 | 46 | 8069188 | ||
BC101686.1 | - | 1 | 46 | 8069192 |
