siRNA Details
Gene Symbol: OR51I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004754.2 | NP_001004754.1 | 1 | 887 | ||
GenBank mRNA | BC140730.1 | - | 1 | 887 | 9021598 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR51I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004754.2 | NP_001004754.1 | 1 | 887 | ||
GenBank mRNA | BC140730.1 | - | 1 | 887 | 9021598 |