siRNA Details
Gene Symbol: OR52N5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001922.2 | NP_001001922.2 | 1 | 741 | ||
GenBank mRNA | BC136979.1 | - | 1 | 741 | 9020971 | |
BC136981.1 | - | 1 | 741 | 9020973 |
