siRNA Details
Gene Symbol: PLCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303012.1 | NP_001289941.1 | 21 | 3514 | ||
XM_011542455.2 | XP_011540757.1 | 22 | 3750 | |||
XM_011542456.2 | XP_011540758.1 | 22 | 3707 | |||
XM_017002870.1 | XP_016858359.1 | 22 | 3739 | |||
XM_017002871.1 | XP_016858360.1 | 22 | 3710 | |||
GenBank mRNA | AB007919.2 | - | 21 | 3514 | ||
AK074149.1 | - | 10 | 1770 | |||
AK122589.1 | - | 19 | 3807 | |||
AK122591.2 | - | 8 | 1653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
