siRNA Details
Gene Symbol: C2orf66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213608.2 | NP_998773.1 | 3 | 1323 | ||
GenBank mRNA | AY358249.1 | - | 3 | 1323 | ||
BC137150.1 | - | 3 | 1199 | 9021147 | ||
BC137151.1 | - | 3 | 1199 | 9021148 |
