siRNA Details
Gene Symbol: FAM166A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001710.2 | NP_001001710.1 | 5 | 814 | ||
XM_006717108.2 | XP_006717171.2 | 6 | 1646 | |||
XM_011518694.1 | XP_011516996.1 | 6 | 1799 | |||
XM_011518695.1 | XP_011516997.1 | 6 | 1685 | |||
XM_017014718.1 | XP_016870207.1 | 5 | 1654 | |||
GenBank mRNA | AK097419.1 | - | 5 | 1034 | ||
AY604177.1 | - | 5 | 792 | |||
BC132916.1 | - | 5 | 759 | 40146938 | ||
BC132918.1 | - | 5 | 759 | 40146940 |
