siRNA Details
Gene Symbol: MIR17HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027349.1 | 4 | 505 | |||
NR_027350.1 | 2 | 4596 | ||||
GenBank mRNA | AK024240.1 | - | 1 | 2319 |
Gene Symbol: MIR18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AB176708.1 | - | LIT3364 oncomir-1 |
2 | 4596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
