siRNA Details
Gene Symbol: ZNF611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161499.1 | NP_001154971.1 | 6 | 3064 | ||
NM_001161500.1 | NP_001154972.1 | 5 | 2964 | |||
NM_001161501.1 | NP_001154973.1 | 5 | 2927 | |||
NM_030972.3 | NP_112234.3 | 7 | 3106 | |||
GenBank mRNA | AK021787.1 | - | 1 | 849 | ||
AK091389.1 | - | 2 | 2644 | |||
AK097434.1 | - | 7 | 3106 | |||
AL834123.1 | - | 1 | 2468 | |||
BC000918.2 | - | 1 | 499 | |||
BC028590.1 | - | 9 | 3348 |
