siRNA Details
Gene Symbol: ZNF611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161499.1 | NP_001154971.1 | 6 | 1499 | ||
NM_001161500.1 | NP_001154972.1 | 5 | 1399 | |||
NM_001161501.1 | NP_001154973.1 | 5 | 1362 | |||
NM_030972.3 | NP_112234.3 | 7 | 1541 | |||
GenBank mRNA | AK091389.1 | - | 2 | 1079 | ||
AK097434.1 | - | 7 | 1541 | |||
AL834123.1 | - | 1 | 903 | |||
BC028590.1 | - | 9 | 1783 | |||
BC136343.1 | - | 5 | 1447 | 9020330 | ||
BC143996.1 | - | 4 | 1296 |
