siRNA Details
Gene Symbol: CT45A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007551.4 | NP_001007552.2 | 3 | 639 | ||
NM_001172288.2 | NP_001165759.2 | 3 | 656 | |||
XM_011531341.2 | XP_011529643.1 | 3 | 521 | |||
GenBank mRNA | BC028711.2 | - | 3 | 639 | 4828274 | |
BC140735.1 | - | 3 | 546 | 9021603 | ||
BC140926.1 | - | 3 | 546 | 9056738 | ||
BC144437.1 | - | 3 | 546 | 9052959 |
Gene Symbol: CT45A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017417.2 | NP_001017417.1 | 3 | 638 | ||
XM_005278141.3 | XP_005278198.1 | 3 | 487 | |||
XM_011531352.1 | XP_011529654.1 | 2 | 408 | |||
GenBank mRNA | AY743709.1 | - | 3 | 639 | ||
BC140913.1 | - | 3 | 538 | 9056725 |
Gene Symbol: CT45A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY743710.1 | - | 3 | 639 |
Gene Symbol: CT45A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017435.1 | NP_001017435.1 | 3 | 639 | ||
XM_006724758.3 | XP_006724821.1 | 3 | 556 | |||
XM_011531340.2 | XP_011529642.1 | 3 | 515 | |||
GenBank mRNA | AK098689.1 | - | 3 | 484 | ||
AY743711.1 | - | 3 | 639 |
Gene Symbol: CT45A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017438.2 | NP_001017438.2 | 3 | 639 | ||
XM_005262425.4 | XP_005262482.2 | 3 | 559 | |||
XM_011531351.1 | XP_011529653.1 | 3 | 490 | |||
GenBank mRNA | AY743714.1 | - | 3 | 639 |
Gene Symbol: CT45A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291543.1 | NP_001278472.1 | 3 | 455 | ||
XM_003960082.4 | XP_003960131.1 | 3 | 542 | |||
XM_011531229.2 | XP_011529531.1 | 3 | 521 | |||
GenBank mRNA | BU540102.1 | - | 3 | 458 |
Gene Symbol: CT45A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BP371111.1 | - | 3 | 484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
