siRNA Details
Gene Symbol: SSX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005636.3 | NP_005627.1 | 4 | 306 | ||
NM_175729.1 | NP_783856.1 | 4 | 306 | |||
XM_017029759.1 | XP_016885248.1 | 4 | 364 | |||
GenBank mRNA | AX740218.1 | - | 3 | 248 | ||
BC005325.1 | - | 4 | 306 | 3961688 | ||
BC137394.1 | - | 3 | 273 | 9021392 | ||
BC137395.1 | - | 3 | 273 | 9021393 | ||
U90841.1 | - | 3 | 248 |
Gene Symbol: SSX4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034832.3 | NP_001030004.1 | 4 | 317 | ||
NM_001040612.2 | NP_001035702.1 | 4 | 317 | |||
XM_017029613.1 | XP_016885102.1 | 4 | 364 | |||
GenBank mRNA | AA312651.1 | - | 4 | 411 | ||
BE866727.1 | - | 4 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
