siRNA Details
Gene Symbol: MTMR9LP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026850.1 | 7 | 2301 | |||
GenBank mRNA | AK096109.1 | - | 6 | 1853 | ||
AL833686.1 | - | 9 | 2601 | |||
BC016171.1 | - | 4 | 1146 | |||
BC041632.1 | - | 4 | 956 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
