siRNA Details
Gene Symbol: FAM131C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182623.2 | NP_872429.2 | 7 | 1497 | ||
GenBank mRNA | AK094085.1 | - | 7 | 1386 | ||
BC016848.1 | - | 7 | 1491 | 3913592 | ||
BC037394.1 | - | 7 | 1465 | 5198517 |
