siRNA Details
Gene Symbol: SVBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199342.3 | NP_955374.1 | 2 | 253 | ||
XM_017001226.1 | XP_016856715.1 | 2 | 256 | |||
GenBank mRNA | AK291356.1 | - | 2 | 148 | ||
BC029427.1 | - | 2 | 117 | 4809776 | ||
BQ712089.1 | - | 2 | 253 |
