siRNA Details
Gene Symbol: TRAPPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011658.3 | NP_001011658.1 | 6 | 1532 | ||
NM_001128835.2 | NP_001122307.2 | 6 | 1526 | |||
NM_014563.5 | NP_055378.1 | 5 | 1390 | |||
XM_011545565.1 | XP_011543867.1 | 7 | 1600 | |||
XM_011545566.2 | XP_011543868.1 | 6 | 1456 | |||
GenBank mRNA | AK055671.1 | - | 1 | 697 |
Gene Symbol: LINC00996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034033.1 | 4 | 1974 | |||
GenBank mRNA | AK097158.1 | - | 4 | 1876 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
