siRNA Details
Gene Symbol: FAM150A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207413.3 | NP_997296.1 | 2 | 415 | ||
GenBank mRNA | AY358213.1 | - | 2 | 418 | ||
BC130641.1 | - | 2 | 362 | 40146672 | ||
BC130643.1 | - | 2 | 362 | 40146674 | ||
BC144534.1 | - | 2 | 362 | 9053061 |
