siRNA Details
Gene Symbol: INGX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002226.3 | 1 | 266 | |||
GenBank mRNA | AF149724.1 | - | 1 | 267 | ||
BC101123.1 | - | 1 | 186 | |||
BC101124.1 | - | 1 | 186 | |||
BC117297.1 | - | 1 | 91 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
