siRNA Details
Gene Symbol: PTRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002913.1 | NP_001002913.1 | 1 | 72 | ||
XM_006716955.3 | XP_006717018.1 | 1 | 1129 | |||
XM_017014276.1 | XP_016869765.1 | 1 | 1129 | |||
XM_017014277.1 | XP_016869766.1 | 1 | 1129 | |||
XM_017014278.1 | XP_016869767.1 | 1 | 1129 | |||
GenBank mRNA | AK090922.1 | - | 2 | 1511 | ||
BC047012.1 | - | 1 | 72 | 5001221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
