siRNA Details
Gene Symbol: CYFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287810.2 | NP_001274739.1 | 5 | 513 | ||
NM_001324119.1 | NP_001311048.1 | 4 | 342 | |||
NM_001324120.1 | NP_001311049.1 | 4 | 521 | |||
NM_001324122.1 | NP_001311051.1 | 4 | 394 | |||
NM_001324123.1 | NP_001311052.1 | 4 | 421 | |||
NM_001324124.1 | NP_001311053.1 | 3 | 304 | |||
NM_001324126.1 | NP_001311055.1 | 4 | 394 | |||
NM_014608.4 | NP_055423.1 | 4 | 394 | |||
XM_011543873.2 | XP_011542175.1 | 4 | 344 | |||
XM_011543874.1 | XP_011542176.1 | 4 | 377 | |||
XM_011543876.2 | XP_011542178.2 | 4 | 350 | |||
XM_017022023.1 | XP_016877512.1 | 4 | 350 | |||
XM_017022024.1 | XP_016877513.1 | 4 | 358 | |||
GenBank mRNA | AK125936.1 | - | 3 | 447 | ||
AK291604.1 | - | 4 | 321 | |||
AY763577.1 | - | 4 | 358 | |||
AY763578.1 | - | 5 | 424 | |||
BC005097.1 | - | 4 | 285 | 3163591 | ||
BF979687.1 | - | 362 | ||||
D38549.1 | - | 4 | 288 | |||
DA317955.1 | - | 4 | 373 | |||
KJ534819.1 | - | 3 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
