siRNA Details
Gene Symbol: ARHGEF37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001669.2 | NP_001001669.2 | 13 | 4431 | ||
XM_005268447.4 | XP_005268504.1 | 13 | 4402 | |||
XM_005268448.1 | XP_005268505.1 | 12 | 4305 | |||
XM_006714784.3 | XP_006714847.1 | 14 | 4770 | |||
XM_011537642.2 | XP_011535944.1 | 13 | 4453 | |||
XM_017009460.1 | XP_016864949.1 | 12 | 4544 | |||
XM_017009461.1 | XP_016864950.1 | 11 | 4194 | |||
GenBank mRNA | AK023814.1 | - | 1 | 3470 | ||
AK123597.1 | - | 5 | 3071 |
