siRNA Details
Gene Symbol: LINC01164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038365.1 | 4 | 2029 | |||
GenBank mRNA | AK128177.1 | - | 4 | 2029 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038365.1 | 4 | 2029 | |||
GenBank mRNA | AK128177.1 | - | 4 | 2029 |