siRNA Details
Gene Symbol: FAM150B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002919.2 | NP_001002919.2 | 2, 3 | 604 | ||
XM_005264689.4 | XP_005264746.1 | 2, 3 | 617 | |||
XM_006711879.3 | XP_006711942.1 | 2, 3 | 618 | |||
XM_011510342.2 | XP_011508644.1 | 2, 3 | 629 | |||
GenBank mRNA | AY358517.1 | - | 2, 3 | 73 | ||
BC033490.1 | - | 2, 3 | 478 |
