siRNA Details
Gene Symbol: ALG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004127.2 | NP_001004127.2 | 3 | 651 | ||
GenBank mRNA | AK025456.1 | - | 3 | 624 | ||
AK128503.1 | - | 2 | 3556 | |||
AK296747.1 | - | 3 | 641 | |||
BC010857.2 | - | 1 | 233 | |||
BC111022.1 | - | 3 | 624 | |||
BC142998.1 | - | 3 | 650 | 8860455 |
