siRNA Details
Gene Symbol: C1orf168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004303.4 | NP_001004303.3 | 8 | 1582 | ||
XM_011540898.1 | XP_011539200.1 | 9 | 1660 | |||
XM_011540899.1 | XP_011539201.1 | 9 | 1660 | |||
XM_011540900.2 | XP_011539202.1 | 9 | 1660 | |||
XM_011540903.2 | XP_011539205.1 | 9 | 1660 | |||
XM_011540904.2 | XP_011539206.1 | 6 | 407 | |||
XM_011540906.2 | XP_011539208.1 | 4 | 175 | |||
XM_017000544.1 | XP_016856033.1 | 9 | 1346 | |||
XM_017000545.1 | XP_016856034.1 | 9 | 1406 | |||
XM_017000546.1 | XP_016856035.1 | 9 | 1660 | |||
XM_017000547.1 | XP_016856036.1 | 9 | 1660 | |||
XM_017000548.1 | XP_016856037.1 | 9 | 1660 | |||
GenBank mRNA | AK093468.1 | - | 4 | 183 | ||
BX648439.1 | - | 8 | 1258 |
