siRNA Details
Gene Symbol: C1orf168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004303.4 | NP_001004303.3 | 20 | 2890 | ||
XM_005270584.3 | XP_005270641.1 | 16 | 2566 | |||
XM_011540898.1 | XP_011539200.1 | 21 | 2968 | |||
XM_011540899.1 | XP_011539201.1 | 19 | 2878 | |||
XM_011540904.2 | XP_011539206.1 | 18 | 1715 | |||
XM_011540906.2 | XP_011539208.1 | 16 | 1483 | |||
XM_017000544.1 | XP_016856033.1 | 21 | 2654 | |||
XM_017000545.1 | XP_016856034.1 | 21 | 2714 | |||
GenBank mRNA | AK093468.1 | - | 14 | 1401 | ||
BX648439.1 | - | 20 | 2568 |
