siRNA Details
Gene Symbol: TMEM220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004313.1 | NP_001004313.1 | 6 | 1056 | ||
XM_005256634.4 | XP_005256691.1 | 5 | 991 | |||
XM_011523835.2 | XP_011522137.1 | 5 | 540 | |||
XM_011523836.2 | XP_011522138.1 | 4 | 932 | |||
GenBank mRNA | AY550194.1 | - | 6 | 1056 | ||
BC136840.1 | - | 6 | 618 | 9020831 |
