siRNA Details
Gene Symbol: ZNF788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027049.1 | 3 | 2899 | |||
GenBank mRNA | AK128282.1 | - | 1 | 1956 | ||
AK128700.1 | - | 3 | 2899 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
