siRNA Details
Gene Symbol: PLEKHG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004330.2 | NP_001004330.1 | 12 | 3208 | ||
GenBank mRNA | AK128530.1 | - | 12 | 3165 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PLEKHG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004330.2 | NP_001004330.1 | 12 | 3208 | ||
GenBank mRNA | AK128530.1 | - | 12 | 3165 |