siRNA Details
Gene Symbol: KIR2DL5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020535.3 | NP_065396.1 | 8 | 1072 | ||
GenBank mRNA | AF204903.1 | - | 8 | 1072 | ||
AF217487.1 | - | 10 | 1041 |
Gene Symbol: KIR2DL5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018081.1 | NP_001018091.1 | 8 | 1017 | ||
GenBank mRNA | AF271607.1 | - | 8 | 1017 | ||
AF271608.1 | - | 8 | 1017 | |||
AJ544587.1 | - | 9 | 1018 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
